Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor® 720 succinimidyl ester
iFluor® 720 succinimidyl ester
iFluor® 720 succinimidyl ester
iFluor® 720 succinimidyl ester
Catalog # 77461-358
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
iFluor® 720 succinimidyl ester
Catalog # 77461-358
Supplier:
AAT BIOQUEST INC
Supplier Number:
1046
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77461-358
Specifications
About this item
Product Details
:
:
1089.29 g/mol
Product Details