Order Entry
United States
ContactUsLinkComponent
Xylene (mixture of isomers), Technical Grade
Flammable
Irritant
Health
Xylene (mixture of isomers), Technical Grade
Catalog # TS444240050
New Product
CAS Number:  
Flammable
Irritant
Health
Xylene (mixture of isomers), Technical Grade
Catalog # TS444240050
Supplier Number:  444240050
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 L
  • Cat. No.:
    TS444240050

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Density:
    0.860 to 0.875 kg/l
  • GC:
    ≥98.5 % (Mixture of isomers)

Specification Test Results

About this item

 :
 :106.17 g/mol
Boiling Pt:136…140 °C (760 mmHg)
Melting Pt:–34 °C
Density:0.865 g/cm³
Flash Pt:23 °C
UN:1307
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264