Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Violet 660 AM
Violet 660 AM, CytoCalcein™ excited at 405 nm
Violet 660 AM, CytoCalcein™ excited at 405 nm
77461-888
:
AAT BIOQUEST INC
:
Violet 660 AM, CytoCalcein™ excited at 405 nm
77461-888
:
AAT BIOQUEST INC
:
21904
:
Size:
10x50 µg
Cat. No.:
77461-888
:
:
812.87 g/mol