Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
CypHer5E NHS ester
CypHer5E NHS ester
CypHer5E NHS ester
CypHer5E NHS ester
Catalog # 77461-892
Supplier:
AAT BIOQUEST INC
CAS Number:
Expand image
CypHer5E NHS ester
Catalog # 77461-892
Supplier:
AAT BIOQUEST INC
Supplier Number:
21912
CAS Number:
Specifications
Size:
1 mg
Cat. No.:
77461-892
Specifications
About this item
Product Details
:
:
827.02 g/mol
Product Details