Order Entry
United States
ContactUsLinkComponent
Eppendorf® Reference® 2 Mechanical Single-Channel Pipettes - Trade-In
Eppendorf® Reference® 2 Mechanical Single-Channel Pipettes - Trade-In
Catalog # 77394-826
Supplier:  Eppendorf
Eppendorf® Reference® 2 Mechanical Single-Channel Pipettes - Trade-In
Catalog # 77394-826
Supplier:  Eppendorf
Supplier Number:  2231302066

Specifications

  • Volume:
    0.1 - 2.5 µl
  • Description:
    Eppendorf® Reference® 2 mechanical single-channel pipette with Spring-loaded tip
  • Cat. No.:
    77394-826
  • Increments:
    0.002 µl
  • No. of channels:
    1
  • Color code:
    Dark grey

Specifications

About this item

Take advantage and trade in ANY brand pipette for a NEW Eppendorf® Reference® 2 single-channel variable-volume Pipette at a reduced price. Trust Eppendorf's premium products for quality in your laboratory, backed by 75 years of liquid handling experience and green, cutting-edge manufacturing.

  • Integrated pipetting system with internal RFID technology
  • Serial number is printed on multiple components of pipette
  • Color coded and volume labeled pipette for quick identification of volume size/tip size
  • Round upper part of pipette makes it possible to work in every position
  • Four-digit display for exact, quick and secure volume setting (volume lock included)
  • Spring-loaded tip cone reduces effort during operation/ejection
  • Sturdy upper handle, movable parts and external edges made from stainless-steel make pipette robust

High flexibility with ability to remove single cones
Ensures continuous identical pipet alignment during work process
Fully autoclavable pipet with smooth surface and few rough edges
Single-button operation
Has Spring-loaded tip cone, movable parts, stainless steel external edges with customizable labeling area
Ergonomic handling with reduced operating effort
Simple and quick tip ejection with active aerosol reduction
Reduce repetitive strain injuries by not having to move your thumb sideways
Haptic feedback before tip ejection
High precision and accuracy provides reliable pipetting results
User friendly secondary adjustment

Air-cushion pipetting system, autoclavable and has epT.I.P.S.® consumables.

Applications: Pipetting of aqueous liquids, single-button operation.