Order Entry
United States
ContactUsLinkComponent
DLG (Dilauroyl-sn-glycerol)
DLG (Dilauroyl-sn-glycerol)
Supplier:  Sino Biological
New Product
Certificates
DLG (Dilauroyl-sn-glycerol)
Supplier:  Sino Biological
New Product

Specifications

  • Storage Conditions:
    For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Formulation:
    Frozen substrate solution containing 10 mM of 1,2-Dilauroyl-sn-glycerol (DLG) in DMSO

Specifications

Related Information

Frequently Bought Together