ContactUsLinkComponent
iFluor® 647 PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
iFluor® 647 PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-528
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 647 PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-528
Supplier:  AAT BIOQUEST INC
Supplier Number:  17043
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    25 nmoles
  • Cat. No.:
    77461-528
Specifications
About this item
This iFluor® 647-PEG12-dUTP conjugate has the same spectral properties as ChromaTide™ Alexa Fluor™ 647-dUTP.
  • PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labeled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
Caution: For research use only.