ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # 77462-178
Supplier: AAT BIOQUEST INC
CAS Number:
Specifications
- Description:ReadiUse™ Universal kinase reaction stopping buffer, 100 ml
- Cat. No.:77462-178
Specifications
About this item
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
- Designed for ease of use
- Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Caution: For research use only.