QUIDEL® QuickVue Rapid COVID-19 Tests
76553-984
:
- Description:QuickVue At-Home OTC COVID-19 test
- Size:25 tests
- Cat. No.:76553-984
The QuickVue At-Home OTC COVID-19 Test (nasal) is intended for the qualitative detection of nucleocapsid proteins from SARS-CoV-2 from individuals with or without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over two or three days with at least 24 hours and no more than 36 hours between tests.