About this item
Unleash your true colours with the BradyJet™ J7300 Colour Label Printer. Create vibrant, industrial-grade inkjet for safety labels, signs and tags in full colour. Non-WiFi option available.
- Full color printing capability
- PC connected
- Print resolution: 4800 dpi
- 2 year warranty
- Maximum label width: 4.25"
- Maximum labels per day: 5000
Ideal for industrial labeling that changes frequently from GHS, marine (BS 5609) and barcoding to slides, tubes, and vials. LabelSense™ technology enables printer, materials, and inks to talk to each other for automatic part recognition, setup, and adjustments. Dual CMY and true black (K) ink cartridges deliver vibrant colours and deeper darks for lab identification that stands out. Prints smear-free inks on labels that resist water, chemicals, and abrasions for up to two-year outdoor durability (even longer indoors). Print Utility dashboard helps users plan, budget, and troubleshoot with status alerts, a job cost calculator, remote monitoring ,and more. Exterior LED lighting and clear cover communicate real-time printer status.
Brady Workstation Laboratory ID Suite makes it easy to create labels that fit any lab through step-by-step templated and custom label creation. Optimized Windows® driver allows connection to third-party software and print PDFs.