About this item
Upgrade the durability and longevity of identification projects, even in the toughest environments, with PermaShield™ Polyester Labels designed for BradyJet printers.
- Permanent acrylic adhesive
- Matte finish
- Durable, chemical-resistant material
- Suitable for harsh environment, outdoor use
- Service temperature range: –40 to +140 °F
PermaShield™ by Brady is a 2-in-1 package that includes a printable label plus matching clear polyester overlaminate. Simply print label, adhere to surface, and apply the matching overlaminate. Protects against harsh solvents, outdoor elements, and food and beverage washdowns. Label topcoat is specially formulated to invigorate colours and keep printed content clear and visible. Commonly used to print general safety signage and labeling. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour Label Printer. Ink cartridges are required to print on this material (use part numbers P-CMY-1 and P-K-1).