Order Entry
United States
ContactUsLinkComponent
General Asymmetrical Dimethylarginine (ADMA) ELISA Kit
General Asymmetrical Dimethylarginine (ADMA) ELISA Kit
Catalog # 77538-404
Supplier:  Bioss
New Product
CAS Number:  
General Asymmetrical Dimethylarginine (ADMA) ELISA Kit
Catalog # 77538-404
Supplier:  Bioss
Supplier Number:  BSKL61341-48T
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    Asymmetrical Dimethylarginine (ADMA)
  • Size:
    1 kit
  • Detection Method:
    Colorimetric microplate reader primary wavelength 450 nm
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 ng/ml
  • Storage Temperature:
    TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at −20 °C.
  • Sensitivity:
    4.63 ng/ml
  • Cat. No.:
    77538-404
  • No. of tests:
    48

Specifications

About this item

  • High sensitivity with a broad assay range
  • Flexible inputs: Serum, plasma, cell culture supernatant
  • Reproducible and specific results