Canine Insulin Like Growth Factor 1 (IGF1) ELISA Kit
Catalog # 77537-020
Supplier: Bioss
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Dog
- Target protein:Insulin Like Growth Factor 1 (IGF1)
- Size:1 kit
- Detection Method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf Life:12 Months
- Detection Range:2.5 - 160 ng/ml
- Storage Temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at −20 °C.
- Sensitivity:0.9 ng/ml
- Cat. No.:77537-020
- No. of tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results