Orders LinkContactUsLinkComponent
Microflex® Mega Texture 93-256 Industrial Gloves
Microflex® Mega Texture 93-256 Industrial Gloves
Supplier:   Ansell Healthcare
Microflex® Mega Texture 93-256 Industrial Gloves
Supplier:  Ansell Healthcare
Certificates
Microflex® Mega Texture 93-256 Industrial Gloves
Microflex® Mega Texture 93-256 Industrial Gloves
5 Options Available Below
Product Details & Documents
Disposable nitrile gloves provide confident grip ​ and durable protection, with a Mega Texture surface for consistent grip in wet and dry conditions.
  • Mega Texture surface for a strong grip
  • Elevated thickness for reliable tear resistance
  • Orange color for high visibility while working
  • 1.5 AQL
Featuring a Mega Texture surface for consistent grip in wet and dry conditions, Microflex® Mega Texture 93-256 gloves have a robust 0.13mm/5.1mil palm thickness for reliable tear resistance. This strong formulation provides durable yet comfortable hand protection, and the high visibility orange finish increases visibility while in use. Ideal for a wide range of food handling applications.
Microflex® Mega Texture 93-256 industrial gloves are recommended for use in automotive, food processing, machinery, equipment, maintenance, manufacturing, recycling, waste disposal, and transportation industries.
Certifications: ASTM D6319. CE 0493. EC 1935/2004, EC 2023/2006. EN ISO 21420:2020, EN ISO 374-1:2016+A1:2018, EN ISO 374-5:2016. FDA21 CFR 177-2600-US Food Contact approved. ISO 13485, ISO 9001, ISO 18889:2019. UKCA 0321.
Packaging: Gloves are packaged 100 per dispenser, 10 dispensers per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 77322-790
  • 77322-792
  • 77322-794
  • 77322-796
  • 77322-798
  • Model:
  • 93-256
  • 93-256
  • 93-256
  • 93-256
  • 93-256
  • Disposable/Reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex/Latex-free:
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Powdered/Powder-free:
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Mega Texture surface
  • Mega Texture surface
  • Mega Texture surface
  • Mega Texture surface
  • Mega Texture surface
  • Cuff Style:
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Anti-Static:
  • +
  • +
  • +
  • +
  • +
Specifications
Product Family Options
Product Information
  • Glove size
    Glove color
    Length
    Availability
    Price
  • Small
    Orange
    240 mm (9.45")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $13.29
    $13.29
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $132.88
    $132.88
    New Customer Promotional Price
    Pack of 100
       $13.29
    Case of 1000
       $132.88
    Supplier Number: 93256070
    Specifications:
    More
    Specifications
    Cat. No.
    77322-790
    Model
    93-256
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Material
    Nitrile
    Finish
    Mega Texture surface
    Cuff Style
    Rolled
    Sterility
    Non sterile
    Anti-Static
    +
  • Medium
    Orange
    240 mm (9.45")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $13.29
    $13.29
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $132.88
    $132.88
    New Customer Promotional Price
    Pack of 100
       $13.29
    Case of 1000
       $132.88
    Supplier Number: 93256080
    Specifications:
    More
    Specifications
    Cat. No.
    77322-792
    Model
    93-256
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Material
    Nitrile
    Finish
    Mega Texture surface
    Cuff Style
    Rolled
    Sterility
    Non sterile
    Anti-Static
    +
  • Large
    Orange
    240 mm (9.45")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $13.29
    $13.29
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $132.88
    $132.88
    New Customer Promotional Price
    Pack of 100
       $13.29
    Case of 1000
       $132.88
    Supplier Number: 93256090
    Specifications:
    More
    Specifications
    Cat. No.
    77322-794
    Model
    93-256
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Material
    Nitrile
    Finish
    Mega Texture surface
    Cuff Style
    Rolled
    Sterility
    Non sterile
    Anti-Static
    +
  • X-Large
    Orange
    240 mm (9.45")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $13.29
    $13.29
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $132.88
    $132.88
    New Customer Promotional Price
    Pack of 100
       $13.29
    Case of 1000
       $132.88
    Supplier Number: 93256100
    Specifications:
    More
    Specifications
    Cat. No.
    77322-796
    Model
    93-256
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Material
    Nitrile
    Finish
    Mega Texture surface
    Cuff Style
    Rolled
    Sterility
    Non sterile
    Anti-Static
    +
  • 2X-Large
    Orange
    240 mm (9.45")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $13.29
    $13.29
    New Customer Promotional Price
    Log in for availability
    Case of 1000
    Case of 1000 - $132.88
    $132.88
    New Customer Promotional Price
    Pack of 100
       $13.29
    Case of 1000
       $132.88
    Supplier Number: 93256110
    Specifications:
    More
    Specifications
    Cat. No.
    77322-798
    Model
    93-256
    Disposable/Reusable
    Disposable
    Latex/Latex-free
    Latex-Free
    Powdered/Powder-free
    Powder-Free
    Material
    Nitrile
    Finish
    Mega Texture surface
    Cuff Style
    Rolled
    Sterility
    Non sterile
    Anti-Static
    +