Human Amyloid Beta Precursor Protein Binding Protein 2 (APPBP2) ELISA Kit
Catalog # 77537-910
Supplier: Bioss
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Amyloid Beta Precursor Protein Binding Protein 2 (APPBP2)
- Size:1 kit
- Detection Method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf Life:12 Months
- Detection Range:0.312 - 20 ng/ml
- Storage Temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:0.133 ng/ml
- Cat. No.:77537-910
- No. of tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results