Bovine Interleukin 1 Alpha (IL1a) ELISA Kit
77536-560
: Bioss
New Product
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Bovine
- Target protein:Interleukin 1 Alpha (IL1a)
- Size:1 kit
- Detection Method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf Life:12 Months
- Detection Range:15.6 - 1000 pg/ml
- Storage Temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:5.8 pg/ml
- Cat. No.:77536-560
- No. of tests:48
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results