Brady® Color Polyester Labels
Supplier: Brady Worldwide
Certificates
About this item
Specialized labels are ideal for easily differentiating vials, PCR and microcentrifuge tubes, and slides.
- Permanent acrylic adhesive
- Gloss finish
- Cold-temperature application; suitable for cryogenic storage
- Recommended ribbon: R6400
Designed to withstand abrasions and exposure to chemicals and extreme temperatures, including immersion in liquid nitrogen and hot water baths. Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces such as glass and polypropylene. Available in six color options with a color bar across the top of the label; pre-sized for multiple applications, some options with matching color vial top label.
Frequently Bought Together