Brady® Aggressive Adhesive Multi-Purpose Clear Polyester Labels
Supplier: Brady Worldwide
Certificates
About this item
Versatile labels are ideal for electronic PCB and component identification, barcode labels, rating plates and solar panel identification.
- Permanent acrylic adhesive
- Recommended ribbon: R6000HF
Durable labels are made from tough, clear polyester that’s chemical, oil, water, and extreme temperature resistant. All labels feature a permanent acrylic adhesive that’s excellent for smooth and rough surfaces alike.
Certifications: UL recognized to UL969, CSA accepted.
Frequently Bought Together