Order Entry
United States
ContactUsLinkComponent
Hazardous Waste Bag
Hazardous Waste Bag
Catalog # 77574-720
Supplier:  GRAINGER
New Product
Hazardous Waste Bag
Catalog # 77574-720
Supplier:  GRAINGER
Supplier Number:  3WNA3
New Product

Specifications

  • Capacity:
    1½ gal.
  • Color:
    Yellow
  • Height:
    18"
  • Material:
    Polyethylene
  • Thickness:
    2 mil
  • Cat. No.:
    77574-720

Specifications

About this item

  • Bottom seal type: Flat
  • Width: 12"
  • Max. load: 8 lbs.
  • Max. temperature: 160 °C