Kinase Assay Buffers I
Supplier: Sino Biological
New Product
Certificates
About this item
Kinase assay buffer I is used for assaying kinase protein.
For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Specifications
- Form:Liquid
- Storage Temperature:–20 °C
- Formulation:Frozen solution of buffer containing: 25 mM MOPS, pH 7.2, 12.5 mM B-glycerol-phosphate, 25 mM MgCl₂, 5 mM EGTA, 2 mM EDTA, 0.25 mM DTT.
Specifications
Frequently Bought Together