Order Entry
United States
ContactUsLinkComponent
Lipid Dilution Buffers II
Lipid Dilution Buffers II
Supplier:  Sino Biological
New Product
Certificates
Lipid Dilution Buffers II
Supplier:  Sino Biological
New Product

Specifications

  • Storage Temperature:
    Store product at –20 °C
  • Formulation:
    Frozen solution of buffer containing: 25 mM HEPES, pH 7.5 and 0.5 mM EGTA

Specifications

Related Information

Frequently Bought Together