Condensers for Rotavapor® Rotary Evaporators
Supplier: Buchi
New Product
Certificates
About this item
Original Büchi glassware guarantees the highest levels of efficiency and safety thanks to outstanding precision in manufacturing. For more than 80 years Büchi has been manufacturing innovative and durable glass parts to ease daily laboratory work.
- High throughput due to advanced design of condensers
- Highly resistant glass provides the highest levels of safety (DURAN® borosilicate glass 3.3)
- Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
- Maximum stability and safety due to Plastic+Glass coating
Outstanding glassware for over 80 years: Experience the highest levels of efficiency and safety with original Büchi glassware crafted with exceptional manufacturing precision perfected over decades. The R-80 includes either a V or C condenser to meet your needs best.
Ordering information: Don't miss out on additional BUCHI quality by ordering a suitable evaporating flask in sizes ranging from 50 to 1000 ml.
Frequently Bought Together