White Shovels
About this item
Sampling Systems shovels are great for a wide range of purposes.
- Made from high quality food grade polypropylene
- Single piece construction - robust and easy to clean
The range of shovels is made from food grade polypropylene and are manufactured as a single piece meaning they are perfect for hygienic industries as they are very easy to clean as well as being very strong.
2 Options Available Below
Product Details & Documents
Sampling Systems shovels are great for a wide range of purposes.
- Made from high quality food grade polypropylene
- Single piece construction - robust and easy to clean
The range of shovels is made from food grade polypropylene and are manufactured as a single piece meaning they are perfect for hygienic industries as they are very easy to clean as well as being very strong.
Product Family Options
Product Information
- ColorLengthAvailabilityPrice
- Supplier Number: 8228P-27
- Supplier Number: 8228P-33