PowderThief Manual Samplers
New Product
Widely used in pharma, the PowderThief is ideal for sampling free-flowing powders.
- World leader in powder sampling
- Suitable for sampling free-flowing powders
- Range of tips available
- Made from 316L stainless steel
The longstanding PowderThief takes samples of free-flowing powders and granules in a wide range of sample volumes. The sampler is easy to use and comes in a wide range of lengths. Simply push the sampler into the powder and push down on the handle to take the sample. Move the body downwards to then close in the sample before removing and emptying the tip.
Frequently Bought Together