Orders LinkContactUsLinkComponent
Sampling Tips for UD Sleeve Sampler
Sampling Tips for UD Sleeve Sampler
Sampling Tips for UD Sleeve Sampler
Certificates
Sampling Tips for UD Sleeve Sampler
Sampling Tips for UD Sleeve Sampler
3 Options Available Below
Product Details & Documents
The UD Sleeve sampler is perfect for small volume powder sampling.
  • Extensively used in the pharmaceutical industry
  • Accurate sampling of free-flowing powders
  • Extension rods are avialble to increase the length of the sampler
The UD Sleeve Sampler is ideal for accurate sampling of small volumes of free-flowing powders. It is made from 316L stainless steel and available with a range of tips, enabling samples sizes between 0,1 and 2,5 ml. Various accessories are available, such as extension rods which anable the operator to extend the length of the sampler.
Product Family Options
Product Information
  • Material
    Volume
    Availability
    Price
  • 77576-286
    Sampling tip for UD sleeve sampler
    Sampling tip for UD sleeve sampler
    Stainless steel 316L
    1.3 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-13
  • 77576-288
    Sampling tip for UD sleeve sampler
    Sampling tip for UD sleeve sampler
    Stainless steel 316L
    1.4 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-14
  • 77576-290
    Sampling tip for UD sleeve sampler
    Sampling tip for UD sleeve sampler
    Stainless steel 316L
    1.5 ml
    Log in for availability
    Log in for availability
    Each
    Each - $145.00
    $145.00
    Each
       $145.00
    Supplier Number: 1500W-15