Cohesive Sampler
Catalog # 77576-356
Supplier: SAMPLING SYSTEMS LTD
New Product
Specifications
- Description:Cohesive sampler
- Material:Stainless steel 316
- Environmentally Preferable:
- Cat. No.:77576-356
Specifications
About this item
This cohesive sampler is designed for powders.
- For taking single point samples
- Handle and four sample cells
- 0,25, 0,50, 0,75, 1,0 ml unit doses
Used throughout the pharmaceutical industry from sticky (or cohesive) powders.
Delivery: A nylon bristle can be ordered separately.
Packaging: In a case.