Orders LinkContactUsLinkComponent
Bailers
Bailers
Bailers
Certificates
Bailers
3 Options Available Below
Product Details & Documents
These bailers are made from high quality 316 stainless steel.
  • Heavy duty construction with satin finish
  • Fully welded handles giving a hygienic crevice free finish
Ordering information: Available in three formats.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • Stainless steel 316
    1000 ml
    285 mm
    170 mm
    Log in for availability
    Log in for availability
    Each
    Each - $114.00
    $114.00
    Each
       $114.00
    Supplier Number: A296-1
  • Stainless steel 316
    2000 ml
    340 mm
    210 mm
    Log in for availability
    Log in for availability
    Each
    Each - $143.00
    $143.00
    Each
       $143.00
    Supplier Number: A296-2
  • Stainless steel 316
    4000 ml
    415 mm
    270 mm
    Log in for availability
    Log in for availability
    Each
    Each - $172.00
    $172.00
    Each
       $172.00
    Supplier Number: A296-4