Order Entry
United States
ContactUsLinkComponent
 
Micro spatula, stainless steel
  77577-608
New Product
undefined
Micro spatula, stainless steel
  77577-608
 :  A100-20
New Product

 

  • Length:
    175 mm
  • Environmentally Preferable:
  • Cat. No.:
    77577-608
  • Blade length:
    40 mm
  • Blade width:
    12 mm

 

 

Perfect for transferring small amounts of powder.

  • Ideal for transferring powders, granules and cream
  • Used for surface sampling and dispensing
  • Crevice free

The range of stainless steel microspatulas are perfect for transferring very small amounts of powders, creams and granules and are ideal for use in pharma, food and chemical companies. The crevice free stainless steel construction means the product can be easily cleaned between uses.