ContactUsLinkComponent
X-Ray and Metal Detectable Pallet Knife and Stirrer, PP, Blue
X-Ray and Metal Detectable Pallet Knife and Stirrer, PP, Blue
Catalog # 77578-204
New Product
undefined
X-Ray and Metal Detectable Pallet Knife and Stirrer, PP, Blue
Catalog # 77578-204
Supplier Number:  M1016B-200
New Product
Specifications
  • Cat. No.:
    77578-204
  • Blade length:
    200 mm
Specifications
About this item
The X-ray and metal detectable pallet knife is ideal for scraping and mixing material. The pallet knives are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Long blade, can be used to stir and mix containers
  • Full batch traceability
  • The 200 mm blade can be used to stir and mix containers
To ensure the cleanliness of the pallet knives are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Certifications: These PP pallet knives conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.