Swabbing Tools and Templates
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
An assortment of tools designed for taking cleaning validation samples.
- Telescopic aluminium rods extend up to 3 m
- Adjustable elbows can adjust to the preferred angle
- Templates are available in both sterile and non-sterile formats
- Available from stock
- Fast delivery
This range of swabbing template kits and swab holders allow users to quickly and conveniently take samples for all cleaning validation needs. Telescopic rods are made from aluminium. The 100×100 mm templates are made from HDPE. The swab holder is made from stainless steel 316L.
Frequently Bought Together