ContactUsLinkComponent
SteriWare® Beakers with Handle
  77576-854
New Product
 : 8049H-2000S
undefined
SteriWare® Beakers with Handle
  77576-854
 :  8049H-2000
New Product
 : 8049H-2000S
Select an Option
129 mm
Select an Option
2000 ml
Select an Option
Disposable jug, sterile
Select an Option
152 mm
Select an Option
77576-854
HDPE, translucent.
  • Individually bagged
  • BSE/TSE-free
  • Full batch traceability
  • Non graduated
  • Available as gamma irradiated sterile (SAL 10-6)
Disposable beakers have a wide base which make them very stable. The handle is rounded to make it comfortable to hold.
Frequently Bought Together