Orders LinkContactUsLinkComponent
SteriWare® Disposable Forceps
SteriWare® Disposable Forceps
   Sustainable Options Available
SteriWare® Disposable Forceps
   Sustainable Options Available
Certificates
SteriWare® Disposable Forceps
SteriWare® Disposable Forceps
2 Options Available Below
Product Details & Documents
These diposable tweezers made from 100% virgin HDPE are ideal for picking up items up to 25 mm. Its special dual action design means it can pick up both small and large items.
  • Made from FDA and EC conforming material
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • Gamma irradiated (SAL 10⁻⁶)
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
 : Manufactured and packed in a class 100000 cleanroom environment. Individually bagged in a heat sealed PE bag.
Product Family Options
Product Information
  • Color
    Length
    Availability
    Price
  • White
    115 mm
    Discontinued
    Supplier Number: 8025H-115S
    Specifications:
    More
    Specifications
    Cat. No.
    77576-712
  • White
    115 mm
    Discontinued
    Supplier Number: 8025H-115
    Specifications:
    More
    Specifications
    Cat. No.
    77576-710
  • White
    115 mm
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $94.91
    $94.91
    Case of 100
       $94.91
    Supplier Number: 8025H-115
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78117-072
    New Product
  • White
    115 mm
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $116.10
    $116.10
    Case of 100
       $116.10
    Supplier Number: 8025H-115S
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    78117-118