Orders LinkContactUsLinkComponent
Single-Point Sampler
Single-Point Sampler
Single-Point Sampler
Certificates
Single-Point Sampler
Single-Point Sampler
2 Options Available Below
Product Details & Documents
Single-point sampler for ice, wax, and similar materials.
Product Family Options
Product Information
  • Material
    Length
    Ø
    Volume
    Availability
    Price
  • 77576-580
    Ice sampler
    Ice sampler
    Stainless steel 316L
    550 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    Each
    Each - $467.00
    $467.00
    Each
       $467.00
    Supplier Number: 6000A-550
  • 77576-578
    Ice sampler
    Ice sampler
    Stainless steel 316L
    1050 mm
    40 mm
    50 ml
    Log in for availability
    Log in for availability
    Each
    Each - $509.00
    $509.00
    Each
       $509.00
    Supplier Number: 6000A-1050