Orders LinkContactUsLinkComponent
MaxiCorer Semi-Solid Samplers
MaxiCorer Semi-Solid Samplers
MaxiCorer Semi-Solid Samplers
Certificates
MaxiCorer Semi-Solid Samplers
MaxiCorer Semi-Solid Samplers
2 Options Available Below
Product Details & Documents
The MaxiCorer can be used for sampling or dispensing small quantities of products. This designed for powders and granules, as well as for semi-solids such as cheese and thick grease in production, quality control or laboratories.
  • Fast and efficent core sampler
  • Long and curved blade construction
  • Hygienic design that can be easily cleaned
Product Family Options
Product Information
  • Material
    Length
    Ø
    Insertion depth
    Availability
    Price
  • 77576-596
    Semi-solid sampler
    Semi-solid sampler
    Stainless steel 316
    300 mm
    25 mm
    200 mm
    Log in for availability
    Log in for availability
    Each
    Each - $141.00
    $141.00
    Each
       $141.00
    Supplier Number: 7010A-01
  • 77576-598
    Semi-solid sampler
    Semi-solid sampler
    Stainless steel 316
    600 mm
    25 mm
    500 mm
    Log in for availability
    Log in for availability
    Each
    Each - $172.00
    $172.00
    Each
       $172.00
    Supplier Number: 7010A-02