Sub Sampler Pro Bomb Sampler
Supplier: SAMPLING SYSTEMS LTD
Jump to:
About this item
Essential for flammable liquids as a heavy-duty sampler.
- Metallic parts manufactured from stainless steel
- Unique double action to sample from a set distance
- Can be plunged until the assembly touches the bottom of the container
Specially designed for the environment throughout the world for both chemical and environmental sampling.
Certifications: Conforms to FDA CFR 177.2600.
2 Options Available Below
Product Details & Documents
Essential for flammable liquids as a heavy-duty sampler.
- Metallic parts manufactured from stainless steel
- Unique double action to sample from a set distance
- Can be plunged until the assembly touches the bottom of the container
Specially designed for the environment throughout the world for both chemical and environmental sampling.
Certifications: Conforms to FDA CFR 177.2600.
Product Family Options
Product Information