Orders LinkContactUsLinkComponent
TeleScoop Sampler for Liquids
TeleScoop Sampler for Liquids
TeleScoop Sampler for Liquids
Certificates
TeleScoop Sampler for Liquids
TeleScoop Sampler for Liquids
4 Options Available Below
Product Details & Documents
These telescopic handles can be used when sampling from overhead gantries or difficult to reach places.
  • Strong telescopic handle
  • Range of volumes and styles of collecting vessels
Suitable for taking samples from liquids and suspensions.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Availability
    Price
  • 77576-020
    Sample chamber
    Sample chamber
    Stainless steel 316
    100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $743.00
    $743.00
    Each
       $743.00
    Supplier Number: 1035P-100
  • 77576-524
    Telescopic rod with PTFE clutch
    Telescopic rod with PTFE clutch
    Stainless steel 316
    80 - 150 cm
    Log in for availability
    Log in for availability
    Each
    Each - $841.00
    $841.00
    Each
       $841.00
    Supplier Number: 5065S-1500
  • 77576-526
    Telescopic rod with PTFE clutch
    Telescopic rod with PTFE clutch
    Stainless steel 316L
    150 - 300 cm
    Log in for availability
    Log in for availability
    Each
    Each - $921.00
    $921.00
    Each
       $921.00
    Supplier Number: 5065S-3000
  • 77576-528
    Telescopic rod with PTFE clutch
    Telescopic rod with PTFE clutch
    Stainless steel 316
    150 - 400 cm
    Log in for availability
    Log in for availability
    Each
    Each - $1,580.00
    $1,580.00
    Each
       $1,580.00
    Supplier Number: 5065S-4500