Mini-Pots, with Lid, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
Made of 316L stainless steel.
- Crevice free interior
- Screw on lid
- Very good chemical and temperature resistance
Multi purpose small volume containers with satin finish.
Silicone O-Ring
Product Information
Frequently Bought Together