SteriWare® Capsule Counting Trays
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
The disposable counting trays are made from robust, shatterproof PS. They are suitable for use with all capsules from size 2 to 00.
- BSE/TSE free
- Suitable for capsules size 2 to size 00
- Manufactured and packed in a cleanroom
- Can be used straight from the bag
Certifications: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Packaging: Individually bagged in a heat-sealed laminate bag.
Frequently Bought Together