Micro Spatulas, Stainless Steel
Supplier: SAMPLING SYSTEMS LTD
New Product
Certificates
About this item
These stainless steel micro spatulas are ideal for working with small amounts of powder. It has a flat blade that can be used to lift or decant small amounts of powder.
- Smooth handle
- Robust design
- Easy to use
They can be also used as a manual stirrer for liquids.
Frequently Bought Together