SteriWare® V Sampling Spatulas with Product Retention
New Product
These V spatulas are ideal for goods in and general powder sampling. The angled tip design keeps the powder in the sampler which is perfect for almost all powders. These spatulas can also be used for harder more viscous materials.
- Robust PS construction
- Available in 3 volumes - 100, 150 and 250 mm blade lengths
- Individually bagged and produced in a cleanroom
- Full batch traceability
2 widths are available. The 10 mm is a narrow design ideal for sampling small amounts from bottles. The 20 mm is the standard design for sampling from sacks, bags, kegs and containers.
: These sampling tools conform to all the latest standards including FDA 177.1640, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch and irradiation certificates are available. Both EU and FDA compliant.
Frequently Bought Together