X-Ray and Metal Detectable Long Handle Scoops
Catalog # 77578-216
Supplier: SAMPLING SYSTEMS LTD
New Product
Specifications
- Sterility:-
- Capacity:100 ml
- Color:Blue
- Environmentally Preferable:
- Cat. No.:77578-216
Specifications
About this item
These long handled metal detectable scoops are perfect for both sampling and dispensing when working with powders and granules.
- Robust PP construction gives excellent chemical resistance to many products
- Supplied Individually bagged and produced in a cleanroom
- Full batch traceability
The long handle and the bright blue colour make the scoops easy to spot if they get left behind. They are both X-ray and metal detectable.
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. The robust nature of these scoops means that they can give many years of service or for more demanding applications they can be used as a single use disposable product straight from the bag.
Certifications: These scoops conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.