Order Entry
United States
ContactUsLinkComponent
Scorpion PharmaScoop® Scoops
Scorpion PharmaScoop® Scoops
Catalog # 77577-886
New Product
undefined
Scorpion PharmaScoop® Scoops
Catalog # 77577-886
Supplier Number:  A346-1500
New Product

Specifications

  • Width:
    155 mm
  • Capacity:
    1500 ml
  • Length:
    280 mm
  • Environmentally Preferable:
  • Cat. No.:
    77577-886

Specifications

About this item

These heavy duty sampling scoops are made of 316 stainless steel with satin finish.

  • Ergonomic handle makes scooping easy when space is tight as well as keeping hands clean
  • Hygienic crevice free construction
  • Radiused corners for easy cleaning

The fully welded construction makes these powder scoops ideal for heavy duty or hygienic applications.