Orders LinkContactUsLinkComponent
Label, Clear
Label, Clear
Pharma labels, Transparent, 95×95 mm
Catalog # 77577-180
Supplier #:  8220C-01
Label, Clear
Catalog # 77577-180
Label, Clear
Label, Clear
Pharma labels, Transparent, 95×95 mm
Price
$95.50
Log in for availability
Each
Product Details & Documents
These labels are made from a clear PP film. They are ideal for sealing bags and sacks.
Clear Labels are transparent so that printing underneath the label can still be easily read.
Certifications: Adhesive conforms to FDA CFR175.105.
Specifications
  • Color:
    Transparent
  • Description:
    Pharma labels
  • Dimensions:
    95×95 mm