Toggle Drums
Supplier: SAMPLING SYSTEMS LTD
Jump to:
About this item
These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
- Splash-tight lid secured by toggle clamps
- Hygienic solid rim
- With two side handles
- Fully welded, crevice free interior
- Finished to a high quality mirror polish
- Removable, easy clean silicone seal
Seal is food grade and meets all the latest FDA compliance standards.
7 Options Available Below
Product Details & Documents
These stainless steel containers from 2 litres up to 300 litres are designed for storing and processing powders and liquids.
- Splash-tight lid secured by toggle clamps
- Hygienic solid rim
- With two side handles
- Fully welded, crevice free interior
- Finished to a high quality mirror polish
- Removable, easy clean silicone seal
Seal is food grade and meets all the latest FDA compliance standards.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- CapacityØHeightAvailabilityPrice
- Supplier Number: A926-2
- Supplier Number: A926-5
- Supplier Number: A926-8
- Supplier Number: A926-10
- Supplier Number: A926-15
- Supplier Number: A926-20
- Supplier Number: A926-300
Accessories
Product Information
- Catalog #AvailabilityPrice
- Drum lid, 316L SS, 470 mm int.Ø of drum
Product Information
- Catalog #AvailabilityPrice
- Drum lid, 316L SS, 395 mm int.Ø of drum