Orders LinkContactUsLinkComponent
Cleaning Brushes for Samplers
Cleaning Brushes for Samplers
Cleaning Brushes for Samplers
Certificates
Cleaning Brushes for Samplers
Cleaning Brushes for Samplers
12 Options Available Below
Product Details & Documents
Brushes with nylon bristles in a twisted stainless steel wire frame and aluminium handle. Ideal for cleaning both stainless steel and plastic samplers.
  • Available in a range of lengths and diameters
Product Family Options
Narrow brush
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • Nylon
    7 - 14 mm
    Wire
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $37.50
    $37.50
    Each
       $37.50
    Supplier Number: 8020N-600
  • Nylon
    7 - 14 mm
    Wire
    1100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $59.30
    $59.30
    Each
       $59.30
    Supplier Number: 8020N-1100
  • Nylon
    7 - 14 mm
    Wire
    1600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $75.10
    $75.10
    Each
       $75.10
    Supplier Number: 8020N-1600
  • Nylon
    7 - 14 mm
    Wire
    2100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $65.70
    $65.70
    Each
       $65.70
    Supplier Number: 8020N-2100
Standard brush
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • Nylon
    15 - 30 mm
    Wire
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $41.20
    $41.20
    Each
       $41.20
    Supplier Number: 8020A-600
  • Nylon
    15 - 30 mm
    Wire
    1100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
    Supplier Number: 8020A-1100
  • Nylon
    15 - 30 mm
    Wire
    1600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $56.30
    $56.30
    Each
       $56.30
    Supplier Number: 8020A-1600
  • Nylon
    15 - 30 mm
    Wire
    2100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $65.70
    $65.70
    Each
       $65.70
    Supplier Number: 8020A-2100
Wide brush
Product Information
  • Bristle material
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • Nylon
    30 - 45 mm
    Wire
    600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $37.50
    $37.50
    Each
       $37.50
    Supplier Number: 8020J-600
  • Nylon
    30 - 45 mm
    Wire
    1100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $46.90
    $46.90
    Each
       $46.90
    Supplier Number: 8020J-1100
  • Nylon
    30 - 45 mm
    Wire
    1600 mm
    Log in for availability
    Log in for availability
    Each
    Each - $56.30
    $56.30
    Each
       $56.30
    Supplier Number: 8020J-1600
  • Nylon
    30 - 45 mm
    Wire
    2100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $65.70
    $65.70
    Each
       $65.70
    Supplier Number: 8020J-2100