Order Entry
United States
ContactUsLinkComponent
Proteinase K
Proteinase K
Catalog # PAV3021
CAS Number:  
undefined
Proteinase K
Catalog # PAV3021
Supplier Number:  V3021
CAS Number:  

Specifications

  • Size:
    100 mg
  • Storage Conditions:
    –30...–10 °C
  • Enzyme Name:
    Proteinase K
  • Cat. No.:
    PAV3021
  • Supplier no.:
    V3021

Specifications

About this item

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride, 50% glycerol.

  • Stable at Room Temperature and Easy to Use
  • Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
  • Retains >80% activity at temperatures up to 60°C
  • No resuspension or thawing before use
  • Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. It cleaves peptide bonds adjacent to the carboxylic group of aliphatic and aromatic amino acids and is useful for general digestion of protein in biological samples. It has been purified to remove RNase and DNase activities. The stability of Proteinase K in urea and SDS and its ability to digest native proteins make it useful for a variety of applications including preparation of chromosomal DNA for pulsed-field gel electrophoresis, protein fingerprinting and removal of nucleases from preparations of DNA and RNA. A typical working concentration for Proteinase K is 50-100ug/ml. Formulation: Proteinase K (PK) Solution is supplied at a concentration of 20mg/ml in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride and 50% glycerol.

The stability of Proteinase K in urea and SDS and its ability to digest native proteins make it useful for a variety of applications including preparation of chromosomal DNA for pulsed-field gel electrophoresis, protein fingerprinting, and removal of nucleases from preparations of DNA and RNA. The recommended reaction buffer is 50 mM Tris-HCl (pH 8.0), 10 mM CaCl2.

It has been purified to remove RNase and DNase activities.