



Potassium bis(trimethylsilyl)amide 14% ∼0.6 mol/L in toluene
Catalog # TCP2730-100ML
Supplier: TCI America
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 ml
- Cat. No.:TCP2730-100ML
Specifications
Specification Test Results
- Appearance:Colourless to Light orange to Yellow clear liquid to cloudy liquid
- Concentration (Neutralization titration):12.0 to 14.0 w/w%
Specification Test Results
About this item
:
EINECS:424-100-2
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:40949-94-8
Formula:C₆H₁₈KNSi₂
MDL Number:MFCD00010330