Order Entry
United States
ContactUsLinkComponent
CUBIC-B Tissue-Clearing Reagent, for Animals
CUBIC-B Tissue-Clearing Reagent, for Animals
  TCT3780-25ML
New Product
 :  
CUBIC-B Tissue-Clearing Reagent, for Animals
  TCT3780-25ML
 :  T3780-25ML
New Product
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Description:
    Tissue-clearing reagent, CUBIC-B for animals
  • Cat. No.:
    TCT3780-25ML
  • Application:
    Clarifier and xylene substitutes

 

 

Specifications
Appearance: Colorless to almost colorless clear liquid
Functionality test: to pass test (remaining fluorescence strength)
Decalcification test: max. 0.3 (590 to 600 nm)