Digital Transmitted Light Microscope, AE2000
Product Details & Documents
The AE2000 digital transmitted light microscope is Motic's new model of inverted microscope equipped with Moticam BMH4000X, providing a flexible optical concept to meet best image quality and robust design for a long lifetime under rough lab conditions. This is the perfect microscope solution for routine cell culture in clinical and pharmaceutical laboratories, also offering best options for university teaching.
- Trinocular head Siedentopf type 45° inclined (light split: ²⁰/₈₀) (AE2000 Inverted)
- Widefield high eyepoint eyepiece 10×/20 mm (paired), focusable (AE2000 Inverted)
- CCIS Plan Achromat objective PL PH4×, PL PH 10×, PL PH 20× (AE2000 Inverted)
- Side facing quadruple nosepiece (AE2000 Inverted)
- Phase slider Ph0, BF, Ph1v (AE2000 Inverted)
- Centering telescope (AE2000 Inverted)
- 1920×1080 HDMI monitor with attachable microscopy camera with 4K (3840×2160) resolution in SD card (Moticam BMH4000X)
- 4K through HDMI (Moticam BMH4000X)
- 20 fps at 3840×2160 through USB connection (Moticam BMH4000X)
- 12 V power adapter, CS ring, Motic 4-dot calibration slide, dust cap, manual, 12 mm focusable lens, 30 mm and 38 mm eyepiece adapters, macro tube, macro calibration card (Moticam BMH4000X)
- Motic Images Plus 3.1 for Windows/macOS (Moticam BMH4000X)
The Moticam BMH4000X is a camera that was designed for ease of use and the ability to output fast live images at 4K (3840×2160) pixels in full HD. Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
Delivery information: Accessories include 0.35× C-Mount camera adapter for ¹/₃" chip sensors. Universal attachable mechanical stage with well plate holders (128×86 mm), 35 mm Petri dish holder, 54 mm Petri dish and slide holder, 65 mm Petri dish and Terasaki holder.
Specifications
- Description:AE2000 digital transmitted light microscope