Trinocular Compound Microscope with Moticam BTI10, Camera Bundle, BA210S
77575-306
: Motic
New Product
- Description:BA210S trinocular compound microscope with Moticam BTI10
- Cat. No.:77575-306
The BA210S biological compound microscope is designed for advanced biology students, teachers, doctors, and lab technicians. Every aspect of these microscopes contributes to making your daily work easier. The ergonomic shape of the stand and the comfortability of the different heads enables fatigue-free viewing for many hours.
- Trinocular head Siedentopf type 30° inclined 360° rotating (Light Split 100:0/20:80) (microscope BA210)
- Widefield high eyepoint eyepiece N-WF10×/20mm (microscope BA210)
- Reversed quadruple revolving nosepiece (microscope BA210)
- CCIS Plan Achromat objectives iASC PL 4×, 10×, 40× S, 100× S-Oil (microscope BA210)
- 3WLED module (microscope BA210)
- 10" LCD Android tablet with 1280×800 screen resolution (Moticam BTI10)
- Connection Wi-Fi (2.4 and 5 GHz), Ethernet (Moticam BTI10)
- WiFi resolution 4 MP (2592×1520 pixels) captured resolution (Moticam BTI10)
- WiFi output (802.1 a/b/g/n/ac supporting both hot-spot mode or using existing WiFi network) (Moticam BTI10)
- Preloaded optimized MotiConnect app (Moticam BTI10)
The Moticam BTI10 unlike other Moticams is combined with a 4.0 MP and a 10" Android tablet that will deliver fast live images up to 30 fps on any trinocular microscope with the right C-mount adapter. With the convenience of a detachable tablet and WiFi image data transfer, the flexibility to capture images and bring your images anywhere has never been so easy.